Learn More
2-Iodonitrobenzene-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Iodonitrobenzene-D4 |
| CAS | 1946843-41-9 |
| Molecular Formula | C6 D4 I N O2 |
| Quantity | 0.25 g |
| Synonym | Benzene-1,2,3,4-d4, 5-iodo-6-nitro- (ACI), 5-Iodo-6-nitrobenzene-1,2,3,4-d4 (ACI), 2-Iodonitrobenzene-D4, 1-Nitro-2-iodobenzene-d4 |
| InChI Formula | InChI=1 S/C6H4INO2/c7-5-3-1-2-4-6(5)8(9)10/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c(I)c1[2 H])[N+](=O)[O-] |
| IUPAC Name | 1,2,3,4-tetradeuterio-5-iodo-6-nitrobenzene |
| Molecular Weight (g/mol) | 253.0306 |
| Formula Weight | 252.9538 g/mol |
| Show More |