Learn More
2-Iodo-3-methylpyridine, 97%
Product Code. 11426890
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426890 5 g 5g
11416890 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11426890 Supplier Thermo Scientific Alfa Aesar Supplier No. L20047.06
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
CAS: 22282-58-2 | C6H6IN | 219.03 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22282-58-2
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.03
MDL Number MFCD04039350
InChI Key PTBOOHAPESUXGV-UHFFFAOYSA-N
PubChem CID 339045
IUPAC Name 2-iodo-3-methylpyridine
SMILES CC1=CC=CN=C1I
Specifications
Boiling Point 80°C (0.3 mmHg)
Refractive Index 1.625
Quantity 5 g
Beilstein 5328169
Sensitivity Light sensitive
Synonym 2-iodo-3-methyl-pyridine, pyridine, 2-iodo-3-methyl, pubchem14399, acmc-1cgxh, pyridine,2-iodo-3-methyl, 2-iodanyl-3-methyl-pyridine, 2-iodo-3-methylpyridine, 2-iodo-3-methyl pyridine
Formula Weight 219.03
Percent Purity 97%
Chemical Name or Material 2-Iodo-3-methylpyridine