Learn More
(+)-2-Hydroxypinocamphone, TRC
Specifications
Specifications
| Chemical Name or Material | (+)-2-Hydroxypinocamphone |
| CAS | 20536-50-9 |
| Molecular Formula | C10H16O2 |
| Quantity | 250 mg |
| Synonym | trans-2-Hydroxypinocamphone, [1R-(1alpha,2beta,5alpha)]-2-Hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one, (1R,2S,5R)-(+)-2-Hydroxy-3-pinanone, (1R,2S,5R)-2-Hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one |
| InChI Formula | InChI=1S/C10H16O2/c1-9(2)6-4-7(9)10(3,12)8(11)5-6/h6-7,12H,4-5H2,1-3H3/t6-,7-,10+/m1/s1 |
| SMILES | CC1(C)[C@@H]2C[C@H]1[C@](C)(O)C(=O)C2 |
| IUPAC Name | (1R,4S,5R)-4-hydroxy-4,6,6-trimethylbicyclo[3.1.1]heptan-3-one |
| Molecular Weight (g/mol) | 168.23 |
| Formula Weight | 168.115 |