Learn More
2-Hydroxy Carbamazepine, TRC
Product Code. 30003362
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003362 1 mg 1mg
30003336 5 mg 5mg
30003513 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30003362 Supplier Toronto Research Chemicals Supplier No. TRCH8841001MG
 Restricted Item
Estimated Shipment: 23-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68011-66-5
Molecular Formula C15 H12 N2 O2
Molecular Weight (g/mol) 252.27
Synonym 2-Hydroxy-5H-dibenz[b,f]azepine-5-carboxamide, 2-Hydroxycarbamazepine, 5H-Dibenz[b,f]azepine-5-carboxamide, 2-hydroxy-
IUPAC Name 3-hydroxybenzo[b][1]benzazepine-11-carboxamide
SMILES NC(=O)N1c2ccccc2C=Cc3cc(O)ccc13
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C15H12N2O2/c16-15(19)17-13-4-2-1-3-10(13)5-6-11-9-12(18)7-8-14(11)17/h1-9,18H,(H2,16,19)
Formula Weight 252.0899
Percent Purity >95%
Chemical Name or Material 2-Hydroxy Carbamazepine
Grade HPLC