Learn More
2-Hydroxy-5-nitrobenzamide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-5-nitrobenzamide |
| CAS | 2912-78-9 |
| Molecular Formula | C7H6N2O4 |
| Quantity | 50 mg |
| Synonym | 5-Nitrosalicylamide |
| InChI Formula | InChI=1S/C7H6N2O4/c8-7(11)5-3-4(9(12)13)1-2-6(5)10/h1-3,10H,(H2,8,11) |
| SMILES | NC(=O)c1cc(ccc1O)[N+](=O)[O-] |
| IUPAC Name | 2-hydroxy-5-nitrobenzamide |
| Molecular Weight (g/mol) | 182.13 |
| Formula Weight | 182.0328 |