missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-5'-methylacetophenone 98%, ACROS Organics™

Product Code. 10703871 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
50g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10703871 50g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10703871 Supplier Thermo Scientific Acros Supplier No. 121380500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1450-72-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 45°C to 49°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H10O2
MDL Number MFCD00002380
Quantity 50 g
Beilstein 08, 111
Synonym 2'-hydroxy-5'-methylacetophenone, 1-2-hydroxy-5-methylphenyl ethanone, 2-hydroxy-5-methylacetophenone, o-acetyl-p-cresol, ethanone, 1-2-hydroxy-5-methylphenyl, 2-acetyl-4-methylphenol, 1-2-hydroxy-5-methylphenyl ethan-1-one, 1-hydroxy-2-acetyl-4-methylbenzene, 1-2-hydroxy-5-methyl-phenyl-ethanone, unii-11661u1zen
InChI Key YNPDFBFVMJNGKZ-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)O)C(=O)C
IUPAC Name 1-(2-hydroxy-5-methylphenyl)ethanone
Molecular Weight (g/mol) 150.177
PubChem CID 15068
Formula Weight 150.18
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2'-Hydroxy-5'-methylacetophenone, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.