Learn More
2-Hydroxy-5-[2-(4-hydroxyphenyl)propan-2-yl]benzoic Acid, TRC
Product Code. 30399768
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30399768 250 mg 250mg
30396402 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30399768 Supplier Toronto Research Chemicals Supplier No. TRCH953975250MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Hydroxy-5-[2-(4-hydroxyphenyl)propan-2-yl]benzoic Acid
CAS 101949-49-9
Molecular Formula C16H16O4
Quantity 250 mg
InChI Formula InChI=1S/C16H16O4/c1-16(2,10-3-6-12(17)7-4-10)11-5-8-14(18)13(9-11)15(19)20/h3-9,17-18H,1-2H3,(H,19,20)
SMILES CC(C)(c1ccc(O)cc1)c2ccc(O)c(c2)C(=O)O
IUPAC Name 2-hydroxy-5-[2-(4-hydroxyphenyl)propan-2-yl]benzoic acid
Molecular Weight (g/mol) 272.3
Formula Weight 272.1049