Learn More
2-Hydroxy-4-methylbenzaldehyde, TRC
Product Code. 30395660
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395660 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30395660 Supplier Toronto Research Chemicals Supplier No. TRCH505020500MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Hydroxy-4-methylbenzaldehyde
CAS 698-27-1
Molecular Formula C8H8O2
Quantity 500 mg
InChI Formula InChI=1S/C8H8O2/c1-6-2-3-7(5-9)8(10)4-6/h2-5,10H,1H3
SMILES Cc1ccc(C=O)c(O)c1
IUPAC Name 2-hydroxy-4-methylbenzaldehyde
Molecular Weight (g/mol) 136.15
Formula Weight 136.0524