Learn More
2-Hydroxy-4-(1-methyl-1-nitroethyl)-tetrahydrofuran(Mixture of Diastereomers), TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-4-(1-methyl-1-nitroethyl)-tetrahydrofuran |
| CAS | 176793-54-7 |
| Molecular Formula | C7 H13 N O4 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C7H13NO4/c1-7(2,8(10)11)5-3-6(9)12-4-5/h5-6,9H,3-4H2,1-2H3 |
| SMILES | CC(C)(C1COC(O)C1)[N+](=O)[O-] |
| IUPAC Name | 4-(2-nitropropan-2-yl)oxolan-2-ol |
| Molecular Weight (g/mol) | 175.18 |
| Formula Weight | 175.0845 |