missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-phenyl-1H-indene-1,3(2H)-dione, TRC

Product Code. 30399614
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399614 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399614 Supplier Toronto Research Chemicals Supplier No. TRCH9495755G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-2-phenyl-1H-indene-1,3(2H)-dione
CAS 3761-46-4
Molecular Formula C15 H10 O3
Quantity 5 g
Synonym 1H-Indene-1,3(2H)-dione, 2-hydroxy-2-phenyl-, 1,3-Indandione, 2-hydroxy-2-phenyl- (7CI,8CI), 2-Hydroxy-2-phenyl-1H-indene-1,3(2H)-dione, 2-Hydroxy-2-phenylindan-1,3-dione
InChI Formula InChI=1S/C15H10O3/c16-13-11-8-4-5-9-12(11)14(17)15(13,18)10-6-2-1-3-7-10/h1-9,18H
SMILES OC1(C(=O)c2ccccc2C1=O)c3ccccc3
IUPAC Name 2-hydroxy-2-phenylindene-1,3-dione
Molecular Weight (g/mol) 238.24
Formula Weight 238.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.