missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazino-4-methylpyridine, TRC

Product Code. 30395666
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395666 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395666 Supplier Toronto Research Chemicals Supplier No. TRCH7145031G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydrazino-4-methylpyridine
CAS 4931-00-4
Molecular Formula C6 H9 N3
Quantity 1 g
InChI Formula InChI=1S/C6H9N3/c1-5-2-3-8-6(4-5)9-7/h2-4H,7H2,1H3,(H,8,9)
SMILES Cc1ccnc(NN)c1
IUPAC Name (4-methylpyridin-2-yl)hydrazine
Molecular Weight (g/mol) 123.156
Formula Weight 123.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.