Learn More
2-Hydrazino-1-methyl-1H-benzimidazole Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydrazino-1-methyl-1H-benzimidazole Hydrochloride |
| CAS | 114804-55-6 |
| Molecular Formula | C8H10N4 . (HCl) |
| Quantity | 5 mg |
| Synonym | 1H-Benzimidazole, 2-hydrazinyl-1-methyl-, hydrochloride (1:1) (ACI), 2H-Benzimidazol-2-one, 1,3-dihydro-1-methyl-, hydrazone, monohydrochloride (9CI), 2-Hydrazinyl-1-methyl-1H-1,3-benzodiazole hydrochloride, 2-Hydrazinyl-1-methyl-1H-benzimidazole hydrochloride, 2-Hydrazino-1-methyl-1H-benzimidazole hydrochloride |
| InChI Formula | InChI=1S/C8H10N4.ClH/c1-12-7-5-3-2-4-6(7)10-8(12)11-9;/h2-5H,9H2,1H3,(H,10,11);1H |
| SMILES | Cl.Cn1c(NN)nc2ccccc12 |
| IUPAC Name | (1-methylbenzimidazol-2-yl)hydrazine;hydrochloride |
| Molecular Weight (g/mol) | 198.65 |
| Formula Weight | 198.0672 |