missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2-Heptanol, TRC
Click to view available options
Quantity:
25 g
250 g
Unit Size:
250g
25g
Specifications
Specifications
| Chemical Name or Material | 2-Heptanol |
| CAS | 543-49-7 |
| Molecular Formula | C7 H16 O |
| Quantity | 25 g |
| Synonym | Heptan-2-ol, 2-Heptanol |
| InChI Formula | InChI=1S/C7H16O/c1-3-4-5-6-7(2)8/h7-8H,3-6H2,1-2H3 |
| SMILES | CCCCCC(C)O |
| IUPAC Name | heptan-2-ol |
| Molecular Weight (g/mol) | 116.2 |
| Formula Weight | 116.1201 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction