Learn More
2-(Formylamino)benzoic Acid (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Formylamino)benzoic Acid |
| CAS | 3342-77-6 |
| Molecular Formula | C8H7NO3 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C8H7NO3/c10-5-9-7-4-2-1-3-6(7)8(11)12/h1-5H,(H,9,10)(H,11,12) |
| SMILES | OC(=O)c1ccccc1NC=O |
| IUPAC Name | 2-formamidobenzoic acid |
| Molecular Weight (g/mol) | 165.15 |
| Formula Weight | 165.0426 |