Learn More
2-Fluorocyclohexanone, 96%
Product Code. 15453347
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453347 250 mg 250mg
11371959 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15453347 Supplier Thermo Scientific Alfa Aesar Supplier No. H26032.MD
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
CAS: 694-82-6 | C6H9FO | 116.135 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 694-82-6
Molecular Formula C6H9FO
Molecular Weight (g/mol) 116.135
MDL Number MFCD01076476
InChI Key VQYOFTVCYSPHPG-UHFFFAOYSA-N
Synonym 2-fluorocyclohexanone, cyclohexanone, 2-fluoro, 2-fluorocylcohexanone, acmc-209o8v, 2-fluoro-cyclohexan-1-one
PubChem CID 11829404
IUPAC Name 2-fluorocyclohexan-1-one
SMILES C1CCC(=O)C(C1)F
Specifications
Density 1.08
Boiling Point 78°C (17 mmHg)
Flash Point 71°C (160°F)
Refractive Index 1.444
Quantity 250 mg
Sensitivity Moisture sensitive
Formula Weight 116.13
Percent Purity 96%
Chemical Name or Material 2-Fluorocyclohexanone