missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester, TRC

Product Code. 30398520
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398520 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398520 Supplier Toronto Research Chemicals Supplier No. TRCF59531850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Fluoro-4-(hydroxymethyl)phenylboronic acid pinacol ester
CAS 1314141-37-1
Molecular Formula C13H18BFO3
Quantity 50 mg
InChI Formula InChI=1S/C13H18BFO3/c1-12(2)13(3,4)18-14(17-12)10-6-5-9(8-16)7-11(10)15/h5-7,16H,8H2,1-4H3
SMILES FC1=CC(=CC=C1B2OC(C)(C)C(O2)(C)C)CO
IUPAC Name (3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol
Molecular Weight (g/mol) 252.09
Formula Weight 252.13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.