Learn More
2-Ethynyl-4-aminotoluene, TRC
Specifications
Specifications
| CAS | 134690-40-7 |
| Molecular Formula | C9H9N |
| Quantity | 25 mg |
| Synonym | 3-Ethynyl-4-methyl-benzenamine |
| InChI Formula | InChI=1S/C9H9N/c1-3-8-6-9(10)5-4-7(8)2/h1,4-6H,10H2,2H3 |
| SMILES | Cc1ccc(N)cc1C#C |
| IUPAC Name | 3-ethynyl-4-methylaniline |
| Molecular Weight (g/mol) | 131.17 |
| Formula Weight | 131.0735 |
| Chemical Name or Material | 2-Ethynyl-4-aminotoluene |