Learn More
2-Ethylpyridine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Ethylpyridine |
| CAS | 100-71-0 |
| Molecular Formula | C7 H9 N |
| Quantity | 5 g |
| Synonym | 2-Ethylpyridine, Pyridine, 2-ethyl-, NSC 964, α-Ethylpyridine |
| InChI Formula | InChI=1S/C7H9N/c1-2-7-5-3-4-6-8-7/h3-6H,2H2,1H3 |
| SMILES | CCc1ccccn1 |
| IUPAC Name | 2-ethylpyridine |
| Molecular Weight (g/mol) | 107.15 |
| Formula Weight | 107.0735 |