Learn More
2-Ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine (cis/trans mixture) (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine |
| CAS | 30223-73-5 |
| Molecular Formula | C20 H23 N |
| Quantity | 5 mg |
| Synonym | Pyrrolidine, 2-ethylidene-1,5-dimethyl-3,3-diphenyl- (6CI, 8CI, 9CI, ACI), 2-Ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine (ACI), 1,5-Dimethyl-3,3-diphenyl-2-ethylidenepyrrolidine, 2-Ethylidene-1,5-dimethyl-3,3-diphenyl-1-pyrrolidine, 2-Ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidene, 2-Ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidone, 2-Ethylidene-3,3-diphenyl-1,5-dimethylpyrrolidine, 2-Ethylidine-1,5-dimethyl-3,3-diphenylpyrrolidine, EDDP, EDDP (methadone metabolite) |
| InChI Formula | InChI=1S/C20H23N/c1-4-19-20(15-16(2)21(19)3,17-11-7-5-8-12-17)18-13-9-6-10-14-18/h4-14,16H,15H2,1-3H3/b19-4- |
| SMILES | C\C=C\1/N(C)C(C)CC1(c2ccccc2)c3ccccc3 |
| IUPAC Name | (2Z)-2-ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine |
| Molecular Weight (g/mol) | 277.4 |
| Formula Weight | 277.183 |