Learn More
(±)-2-Ethylhexanoic-d15 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Ethylhexanoic Acid-D15 |
| CAS | 352431-38-0 |
| Molecular Formula | C8 D15 H O2 |
| Quantity | 0.5 g |
| Synonym | Hexanoic-2,3,3,4,4,5,5,6,6,6-d10 acid, 2-(ethyl-d5)- (9 CI), 2-(Ethyl-d5)hexanoic-2,3,3,4,4,5,5,6,6,6-d10 acid, (±)-2-Ethylhexanoic-d15 Acid, 2-Ethylhexanoic Acid-D15 |
| InChI Formula | InChI=1 S/C8H16O2/c1-3-5-6-7(4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)/i1D3,2D3,3D2,4D2,5D2,6D2,7 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C(=O)O)C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,3,4,4,5,5,6,6,6-decadeuterio-2-(1,1,2,2,2-pentadeuterioethyl)hexanoic acid |
| Molecular Weight (g/mol) | 159.3039 |
| Formula Weight | 159.2092 g/mol |
| Show More |