Learn More
2-Ethyl-m-xylene, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethyl-m-xylene |
| CAS | 2870-04-4 |
| Molecular Formula | C10 H14 |
| Quantity | 100 mg |
| Synonym | Benzene, 2-ethyl-1,3-dimethyl-, m-Xylene, 2-ethyl- (7CI,8CI), 2-Ethyl-1,3-dimethylbenzene, 1,3-Dimethyl-2-ethylbenzene, 1-Ethyl-2,6-dimethylbenzene, 2,6-Dimethyl-1-ethylbenzene, 2-Ethyl-m-xylene |
| InChI Formula | InChI=1S/C10H14/c1-4-10-8(2)6-5-7-9(10)3/h5-7H,4H2,1-3H3 |
| SMILES | CCc1c(C)cccc1C |
| IUPAC Name | 2-ethyl-1,3-dimethylbenzene |
| Molecular Weight (g/mol) | 134.22 |
| Formula Weight | 134.1096 |