Learn More
2-Ethyl-d5-toluene, CDN
Product Code. 30321390
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321390 0.25 g 0.25g
30315110 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321390 Supplier CDN Supplier No. CDND78640.25G
 Restricted Item
Estimated Shipment: 10-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Ethyltoluene-D5 (ethyl-D5)
CAS 2982920-37-4
Molecular Formula C9 D5 H7
Quantity 0.25 g
Synonym 1-(Ethyl-1,1,2,2,2-d5)-2-methylbenzene, 2-Ethyltoluene-d5, 2-Ethyltoluene-D5 (ethyl-D5), 2-(Ethyl-d5)toluene, 2-Ethyl-d5-toluene
InChI Formula InChI=1 S/C9H12/c1-3-9-7-5-4-6-8(9)2/h4-7 H,3H2,1-2H3/i1D3,3D2
SMILES [2 H]C([2 H])([2 H])C([2 H])([2 H])c1ccccc1C
IUPAC Name 1-methyl-2-(1,1,2,2,2-pentadeuterioethyl)benzene
Molecular Weight (g/mol) 125.2224
Formula Weight 125.1253 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity