missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethyl-d5-toluene, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Ethyltoluene-D5 (ethyl-D5) |
| CAS | 2982920-37-4 |
| Molecular Formula | C9 D5 H7 |
| Quantity | 0.1 g |
| Synonym | 1-(Ethyl-1,1,2,2,2-d5)-2-methylbenzene, 2-Ethyltoluene-d5, 2-Ethyltoluene-D5 (ethyl-D5), 2-(Ethyl-d5)toluene, 2-Ethyl-d5-toluene |
| InChI Formula | InChI=1 S/C9H12/c1-3-9-7-5-4-6-8(9)2/h4-7 H,3H2,1-2H3/i1D3,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])c1ccccc1C |
| IUPAC Name | 1-methyl-2-(1,1,2,2,2-pentadeuterioethyl)benzene |
| Molecular Weight (g/mol) | 125.2224 |
| Formula Weight | 125.1253 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction