missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethyl-1,1-d2-5,5-dimethyl-1,3-dioxane, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | 2-Ethyl-5,5-dimethyl-1,3-dioxane-D2 (ethyl-1,1-D2) |
| Molecular Formula | C8 D2 H14 O2 |
| Quantity | 0.05 g |
| Synonym | 2-Ethyl-1,1-d2-5,5-dimethyl-1,3-dioxane, 2-(Ethyl-1,1-d2)-5,5-dimethyl-1,3-dioxane, 5,5-Dimethyl-2-(ethyl-1,1-d2)-1,3-dioxane, 2-Ethyl-5,5-dimethyl-1,3-dioxane-D2 (ethyl-1,1-D2) |
| InChI Formula | InChI=1 S/C8H16O2/c1-4-7-9-5-8(2,3)6-10-7/h7H,4-6H2,1-3H3/i4D2 |
| SMILES | [2 H]C([2 H])(C)C1OCC(C)(C)CO1 |
| IUPAC Name | 2-(1,1-dideuterioethyl)-5,5-dimethyl-1,3-dioxane |
| Molecular Weight (g/mol) | 146.2238 |
| Formula Weight | 146.1276 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction