missing translation for 'onlineSavingsMsg'
Learn More

2-Ethenyl-5-oxo-1-pyrrolidinecarboxylic Acid tert-Butyl Ester, TRC

Product Code. 30370372
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370372 500 mg
30386765 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30370372 Supplier Toronto Research Chemicals Supplier No. TRCE890375500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160428-64-8
Molecular Formula C11H17NO3
Molecular Weight (g/mol) 211.26
Synonym 2-Ethenyl-5-oxo-1-pyrrolidinecarboxylic Acid 1,1-Dimethylethyl Ester
IUPAC Name tert-butyl 2-oxo-5-vinylpyrrolidine-1-carboxylate
SMILES C=CC1N(C(OC(C)(C)C)=O)C(CC1)=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H17NO3/c1-5-8-6-7-9(13)12(8)10(14)15-11(2,3)4/h5,8H,1,6-7H2,2-4H3
Formula Weight 211.12
Chemical Name or Material 2-Ethenyl-5-oxo-1-pyrrolidinecarboxylic Acid tert-Butyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.