Learn More
2-Diphenylphosphino-1-naphthoic acid, 97%, Thermo Scientific Chemicals
Product Code. 11416096
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416096 1 g 1g
11426096 5 g 5g
11436096 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11416096 Supplier Thermo Scientific Alfa Aesar Supplier No. B22361.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
CAS 178176-80-2
Molecular Formula C23H17O2P
Molecular Weight (g/mol) 356.36
MDL Number MFCD02683090
InChI Key WJCBDBBZIGUHOB-UHFFFAOYSA-N
Synonym 2-diphenylphosphino-1-naphthoic acid, 2-diphenylphosphanyl naphthalene-1-carboxylic acid, acmc-20anx1, 2-diphenylphosphino-1-naphthalenecarboxylic acid, 2-diphenylphosphino-naphthalen-1-carboxylic acid, 1-naphthalenecarboxylicacid, 2-diphenylphosphino
PubChem CID 10861198
IUPAC Name 2-diphenylphosphanylnaphthalene-1-carboxylic acid
SMILES OC(=O)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1
Especificaciones
Melting Point 190°C to 192°C
Quantity 1 g
Formula Weight 356.36
Percent Purity 97%
Chemical Name or Material 2-Diphenylphosphino-1-naphthoic acid
RUO – Research Use Only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.