missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)-N-methylethylamine, TRC

Product Code. 30388591
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388591 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388591 Supplier Toronto Research Chemicals Supplier No. TRCD91226550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Diethylamino)-N-methylethylamine
CAS 104-79-0
Molecular Formula C7 H18 N2
Quantity 50 mg
Synonym 1,2-Ethanediamine, N1,N1-diethyl-N2-methyl-
InChI Formula InChI=1S/C7H18N2/c1-4-9(5-2)7-6-8-3/h8H,4-7H2,1-3H3
SMILES CCN(CC)CCNC
IUPAC Name N',N'-diethyl-N-methylethane-1,2-diamine
Molecular Weight (g/mol) 130.231
Formula Weight 130.147
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.