missing translation for 'onlineSavingsMsg'
Learn More

2-Descyano-2-ethyl Formyl-tofacitinib, TRC

Product Code. 30134758
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134758 500 mg 500mg
30135273 100 mg 100mg
30011469 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134758 Supplier Toronto Research Chemicals Supplier No. TRCD221590500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1675248-18-6
Molecular Formula C18 H25 N5 O3
Molecular Weight (g/mol) 359.423
Synonym Ethyl 3-((3R,4R)-4-Methyl-3-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)-3-oxopropanoate
IUPAC Name ethyl 3-[(3R,4R)-4-methyl-3-[methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]piperidin-1-yl]-3-oxopropanoate
SMILES CCOC(=O)CC(=O)N1CC[C@@H](C)[C@H](C1)N(C)c2ncnc3[nH]ccc23

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H25N5O3/c1-4-26-16(25)9-15(24)23-8-6-12(2)14(10-23)22(3)18-13-5-7-19-17(13)20-11-21-18/h5,7,11-12,14H,4,6,8-10H2,1-3H3,(H,19,20,21)/t12-,14+/m1/s1
Formula Weight 359.196
Chemical Name or Material 2-Descyano-2-ethyl Formyl-tofacitinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.