missing translation for 'onlineSavingsMsg'
Learn More

2’-Deoxyuridine-2-13C,1,3-15N2, TRC

Product Code. 30351676
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351676 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351676 Supplier Toronto Research Chemicals Supplier No. TRCD28801225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2'-Deoxyuridine-13C,15N2
CAS 369656-76-8
Molecular Formula 13C C8 H12 15N2 O5
Quantity 25 mg
Synonym Uridine-2-13C-1,3-15N2, 2'-deoxy- (9CI), 2'-Deoxyuridine-13C,15N2, 2'-Deoxyuridine-2-13C,1,3-15N2, 2'-Deoxyuridine-2-13C-1,3-15N2
InChI Formula InChI=1S/C9H12N2O5/c12-4-6-5(13)3-8(16-6)11-2-1-7(14)10-9(11)15/h1-2,5-6,8,12-13H,3-4H2,(H,10,14,15)/t5-,6+,8+/m0/s1/i9+1,10+1,11+1
SMILES OC[C@H]1O[C@H](C[C@@H]1O)[15N]2C=CC(=O)[15NH][13C]2=O
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl](2^{13}C,1,3-^{15}N_{2})pyrimidine-2,4-dione
Molecular Weight (g/mol) 231.18
Formula Weight 231.072
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.