Learn More
2-Decyldodecanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Decyldodecanoic Acid |
| CAS | 2874-72-8 |
| Molecular Formula | C22 H44 O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C22H44O2/c1-3-5-7-9-11-13-15-17-19-21(22(23)24)20-18-16-14-12-10-8-6-4-2/h21H,3-20H2,1-2H3,(H,23,24) |
| SMILES | CCCCCCCCCCC(CCCCCCCCCC)C(=O)O |
| IUPAC Name | 2-decyldodecanoic acid |
| Molecular Weight (g/mol) | 340.584 |
| Formula Weight | 340.334 |