Learn More
2-(Cyclopropylmethoxy)pyridine-3-carboxaldehyde, 97%
Product Code. 11424520
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424520 5 g 5g
11414520 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11424520 Supplier Thermo Scientific Alfa Aesar Supplier No. H50091.06
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
CAS: 902835-88-5 | C10H11NO2 | 177.203 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 902835-88-5
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD07781136
InChI Key AEOXNPSSZSDLDK-UHFFFAOYSA-N
Synonym 2-cyclopropylmethoxy nicotinaldehyde, 2-cyclopropylmethoxy pyridine-3-carbaldehyde, 2-cyclopropylmethoxy pyridine-3-carboxaldehyde, 2-cyclopropylmethoxy-pyridine-3-carbaldehyde, 2-cyclopropyl-methoxy pyridine-3-carbaldehyde
PubChem CID 42553129
IUPAC Name 2-(cyclopropylmethoxy)pyridine-3-carbaldehyde
SMILES C1CC1COC2=C(C=CC=N2)C=O
Specifications
Quantity 5 g
Formula Weight 177.2
Percent Purity 97%
Chemical Name or Material 2-(Cyclopropylmethoxy)pyridine-3-carboxaldehyde