Learn More
2-Cyclopentylcyclopentanone, 99%, Thermo Scientific™
Product Code. 11373196
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11373196
Brand: Thermo Scientific Alfa Aesar L12503.14
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4884-24-6
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00051419
InChI Key CWZGKTMWPFTJCS-UHFFFAOYNA-N
Synonym 2-cyclopentylcyclopentanone, 1,1'-bicyclopentyl-2-one, bicyclopentyl-2-one, 1,1'-bi cyclopentan-2-one, bicyclopentylone, acmc-1aqra, 2-cyclopentyl cyclopentanone, cyclopentanone, 2-cyclopentyl
PubChem CID 21003
IUPAC Name 2-cyclopentylcyclopentan-1-one
SMILES O=C1CCCC1C1CCCC1
Specifications
Density 0.98
Boiling Point 230°C to 233°C
Flash Point 104°C (219°F)
Refractive Index 1.478
Quantity 25 g
Beilstein 1306869
Formula Weight 152.24
Percent Purity 99%
Chemical Name or Material 2-Cyclopentylcyclopentanone