Learn More
2-Cyano-3-(3-chlorophenylethyl)pyridine, TRC
Product Code. 30359404
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359404 1 g 1g
30350839 50 mg 50mg
30359278 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30359404 Supplier Toronto Research Chemicals Supplier No. TRCC9624501G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 31255-57-9
Molecular Formula C14 H11 Cl N2
Molecular Weight (g/mol) 242.7
Synonym Loratadine Cyano Impurity
IUPAC Name 3-[2-(3-chlorophenyl)ethyl]pyridine-2-carbonitrile
SMILES Clc1cccc(CCc2cccnc2C#N)c1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C14H11ClN2/c15-13-5-1-3-11(9-13)6-7-12-4-2-8-17-14(12)10-16/h1-5,8-9H,6-7H2
Formula Weight 242.0611
Chemical Name or Material 2-Cyano-3-(3-chlorophenylethyl)pyridine