missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 2-Cl-IB-MECA

Product Code. p-4783162
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001195 10 mg 10mg
18034562 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001195 Supplier Tocris Bioscience™ Supplier No. 1104/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly selective A3 agonist

High affinity and extremely selective A3 adenosine receptor agonist (Ki = 0.33 nM). Displays 2500- and 1400-fold selectivity over A1 and A2A receptors respectively. Exhibits high selectivity over the Na+-independent adenosine transporter.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163042-96-4
Molecular Formula C18H18ClIN6O4

Specifications

Quantity 10 mg
Formula Weight 544.74
Purity >98%
Molecular Weight (g/mol) 544.734
Chemical Name or Material 2-Cl-IB-MECA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.