Learn More
2-Chloropropane-2-d1, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Chloropropane-2-D |
| CAS | 53778-42-0 |
| Molecular Formula | C3 D H6 Cl |
| Quantity | 0.5 g |
| Synonym | Propane-2-d, 2-chloro- (7 CI, 9 CI), 2-Chloro-2-deuteriopropane, 2-Deutero-2-chloropropane, 2-Chloropropane-2-d1, 2-Chloropropane-2-d, 2-Chloropropane-2-D |
| InChI Formula | InChI=1 S/C3H7Cl/c1-3(2)4/h3H,1-2H3/i3D |
| SMILES | [2 H]C(C)(C)Cl |
| IUPAC Name | 2-chloro-2-deuteriopropane |
| Molecular Weight (g/mol) | 79.5468 |
| Formula Weight | 79.0299 g/mol |
| Show More |