missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenylacetic acid, 98%

Product Code. 11472518
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11472518

Brand: Thermo Scientific Alfa Aesar A15834.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2444-36-2 | C8H7ClO2 | 170.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2444-36-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00004317
InChI Key IUJAAIZKRJJZGQ-UHFFFAOYSA-N
Synonym 2-chlorophenylacetic acid, o-chlorophenylacetic acid, 2-2-chlorophenyl acetic acid, 2-chlorophenyl acetic acid, 2-chlorophenylaceticacid, benzeneacetic acid, 2-chloro, o-chlorophenyl acetic acid, acetic acid, o-chlorophenyl, 2-chloro-benzeneacetic acid
PubChem CID 17124
IUPAC Name 2-(2-chlorophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)Cl

Specifications

Melting Point 94°C to 97°C
Boiling Point 145°C (5 mmHg)
Quantity 50 g
Odor Odorless
Beilstein 1101523
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 2-Chlorophenylacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.