missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethoxy)acetic Acid Ethyl Ester, TRC

Product Code. 30360908
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360908 100 mg 100mg
30348934 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30360908 Supplier Toronto Research Chemicals Supplier No. TRCC367815100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124186-08-9
Molecular Formula C5H9ClO3
Molecular Weight (g/mol) 152.58
Synonym Ethyl 2-(Chloromethoxy)acetate
IUPAC Name ethyl 2-(chloromethoxy)acetate
SMILES CCOC(=O)COCCl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H9ClO3/c1-2-9-5(7)3-8-4-6/h2-4H2,1H3
Formula Weight 152.024
Chemical Name or Material 2-(Chloromethoxy)acetic Acid Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.