Learn More
2-Chlorobenzaldehyde Oxime, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chlorobenzaldehyde Oxime |
| CAS | 3717-28-0 |
| Molecular Formula | C7H6ClNO |
| Quantity | 50 g |
| Synonym | 2-Chlorobenzaldehyde oxime, 2-Chlorobenzaldoxime, NSC 61415, o-Chlorobenzaldehyde oxime, o-Chlorobenzaldoxime |
| InChI Formula | InChI=1S/C7H6ClNO/c8-7-4-2-1-3-6(7)5-9-10/h1-5,10H |
| SMILES | ON=Cc1ccccc1Cl |
| IUPAC Name | N-[(2-chlorophenyl)methylidene]hydroxylamine |
| Molecular Weight (g/mol) | 155.58 |
| Formula Weight | 155.0138 |