missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloroacetanilide, 98+%

Product Code. 11427736
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427736 500 g 500g
11407736 25 g 25g
11417736 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427736 Supplier Thermo Scientific Alfa Aesar Supplier No. A14711.36
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 533-17-5 | C8H8ClNO | 169.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-17-5
Molecular Formula C8H8ClNO
Molecular Weight (g/mol) 169.61
MDL Number MFCD00045169
InChI Key KNVQTRVKSOEHPU-UHFFFAOYSA-N
Synonym 2'-chloroacetanilide, n-2-chlorophenyl acetamide, o-chloroacetanilide, acetamide, n-2-chlorophenyl, 2'-chloro acetanilide, acetanilide, 2'-chloro, unii-20c42pa69y, a-chloroacetanilide, acetic acid, amide, n-2-chlorophenyl, acmc-209l5z
PubChem CID 10777
ChEBI CHEBI:35087
IUPAC Name N-(2-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1Cl

Specifications

Melting Point 86°C to 88°C
Quantity 500 g
Beilstein 2086967
Merck Index 14,2111
Formula Weight 169.61
Percent Purity ≥98%
Chemical Name or Material 2'-Chloroacetanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.