missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-p-phenylenediamine sulfate, 95%, Thermo Scientific™

Product Code. 11466946
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466946 1000 g 1000g
11476946 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466946 Supplier Thermo Scientific Alfa Aesar Supplier No. B24029.0B
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61702-44-1
Molecular Formula C6H9ClN2O4S
Molecular Weight (g/mol) 240.66
MDL Number MFCD00013004
InChI Key GQFGHCRXPLROOF-UHFFFAOYSA-N
Synonym 2-chloro-p-phenylenediamine sulfate, 2-chlorobenzene-1,4-diamine sulfate, 2-chloro-1,4-phenylenediamine sulfate, fourrine so, 2-chloro-1,4-benzenediamine sulfate, 2-chlorobenzene-1,4-diamine; sulfuric acid, 1,4-benzenediamine, 2-chloro-, sulfate, renal so, fourrine 81, 2-cl-p-pd
PubChem CID 22584
ChEBI CHEBI:76597
IUPAC Name 2-chlorobenzene-1,4-diamine;sulfuric acid
SMILES OS(O)(=O)=O.NC1=CC=C(N)C(Cl)=C1

Specifications

Melting Point 251°C to 253°C
Quantity 1000 g
UN Number UN2811
Beilstein 8441373
Formula Weight 240.67
Percent Purity 95%
Chemical Name or Material 2-Chloro-p-phenylenediamine sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.