Learn More
2-Chloro-5-nitroanisole, TRC
Product Code. 30356095
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30356095 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30356095 Supplier Toronto Research Chemicals Supplier No. TRCC3737755G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloro-5-nitroanisole
CAS 1009-36-5
Molecular Formula C7 H6 Cl N O3
Quantity 5 g
InChI Formula InChI=1S/C7H6ClNO3/c1-12-7-4-5(9(10)11)2-3-6(7)8/h2-4H,1H3
SMILES COc1cc(ccc1Cl)[N+](=O)[O-]
IUPAC Name 1-chloro-2-methoxy-4-nitrobenzene
Molecular Weight (g/mol) 187.58
Formula Weight 187.0036