Learn More
2-Chloro-5-(chloromethyl)pyridine, TRC
Product Code. 30366266
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366266 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30366266 Supplier Toronto Research Chemicals Supplier No. TRCC3647155G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloro-5-(chloromethyl)pyridine
CAS 70258-18-3
Molecular Formula C6 H5 Cl2 N
Quantity 5 g
Synonym 2-Chloro-5-(chloromethyl)pyridine, (6-Chloro-3-pyridyl)methyl chloride, 2-Chloro-5-pyridylmethyl chloride, 2-Chloropyridin-5-ylmethyl chloride, 3-(Chloromethyl)-6-chloropyridine, 5-(Chloromethyl)-2-chloropyridine, 6-Chloro-3-(chloromethyl)pyridine
InChI Formula InChI=1S/C6H5Cl2N/c7-3-5-1-2-6(8)9-4-5/h1-2,4H,3H2
SMILES ClCc1ccc(Cl)nc1
IUPAC Name 2-chloro-5-(chloromethyl)pyridine
Molecular Weight (g/mol) 162.02
Formula Weight 160.9799