missing translation for 'onlineSavingsMsg'
Learn More

(2-Chloro-4-pyridinyl)methanol, 97%, Thermo Scientific™

Product Code. 10092264
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10092264

Brand: Thermo Scientific Maybridge CC53609DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 100704-10-7
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD06858767
InChI Key UDDVPFLXGOBESH-UHFFFAOYSA-N
Synonym 2-chloropyridin-4-yl methanol, 2-chloropyridine-4-methanol, 2-chloro-4-hydroxymethyl pyridine, 2-chloro-pyridine-4-yl-methanol, 2-chloro-pyridin-4-yl-methanol, 2-chloro-4-pyridinyl methanol, 4-pyridinemethanol, 2-chloro, 2-chloro-4-hydroxymethylpyridine, 2-chloro-4-pyridinemethanol, 2-chloro-4-pyridyl methan-1-ol
PubChem CID 7062237
IUPAC Name (2-chloropyridin-4-yl)methanol
SMILES C1=CN=C(C=C1CO)Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 143.57
Percent Purity 95%
Chemical Name or Material (2-Chloro-4-pyridinyl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.