Learn More
2-Chloro-4-fluorophenylacetonitrile, 97%
Product Code. 11426356
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11426356
Brand: Thermo Scientific Alfa Aesar B23132.14
Request Bulk or Custom Format
CAS: 75279-56-0 | C8H5ClFN | 169.583 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75279-56-0
Molecular Formula C8H5ClFN
Molecular Weight (g/mol) 169.583
MDL Number MFCD00236029
InChI Key GSMCLMKFBYLWRP-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorophenylacetonitrile, 2-chloro-4-fluorobenzyl cyanide, 2-2-chloro-4-fluorophenyl acetonitrile, 2-chloro-4-fluorobenzylcyanide, benzeneacetonitrile, 2-chloro-4-fluoro, 2-chloro-fluorophenylacetonitrile, 2-chloro-4-fluorophenyl acetonitrile, 2-2-chloro-4-fluorophenyl ethanenitrile, pubchem4251
PubChem CID 2725063
IUPAC Name 2-(2-chloro-4-fluorophenyl)acetonitrile
SMILES C1=CC(=C(C=C1F)Cl)CC#N
Specifications
Melting Point 67°C to 69°C
Quantity 25 g
UN Number UN3439
Formula Weight 169.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorophenylacetonitrile