Learn More
2-Chloro-3-methylaniline, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-3-methylaniline |
| CAS | 29027-17-6 |
| Molecular Formula | C7 H8 Cl N |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C7H8ClN/c1-5-3-2-4-6(9)7(5)8/h2-4H,9H2,1H3 |
| SMILES | Cc1cccc(N)c1Cl |
| IUPAC Name | 2-chloro-3-methyl-aniline |
| Molecular Weight (g/mol) | 141.6 |
| Formula Weight | 141.0345 |