Learn More
2-Chloro-3-iodopyridine, 98+%, Thermo Scientific™
Product Code. 10341593 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Product Code. 10341593
Brand: Thermo Scientific Acros 379510050
Request Bulk or Custom Format
Specifications
CAS 78607-36-0
CAS Min % 98.0
CAS Max % 100.0
Melting Point 93°C to 96°C
Color Cream-Yellow to Salmon
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Molecular Formula C5H3ClIN
MDL Number MFCD00661298
Quantity 5 g
Synonym 2-chloro-3-iodo-pyridine, pyridine, 2-chloro-3-iodo, 3-iodo-2-chloropyridine, zlchem 306, pubchem1188, acmc-209pes, 2-chloro-3-iodopyridine, ksc378k0b, 2-chloranyl-3-iodanyl-pyridine, 2-choro-3-iodopyridine
Solubility Information Solubility in water: insoluble
InChI Key OHWSWGXNZDSHLM-UHFFFAOYSA-N
SMILES C1=CC(=C(N=C1)Cl)I
IUPAC Name 2-chloro-3-iodopyridine
Molecular Weight (g/mol) 239.44
PubChem CID 2735743
Formula Weight 239.44
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-iodopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-3-iodopyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.