Learn More
2-Chloro-3-iodopyridine, 97%, Thermo Scientific™
Product Code. 11394426
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11394426
Brand: Thermo Scientific Alfa Aesar L20028.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 78607-36-0
Molecular Formula C5H3ClIN
Molecular Weight (g/mol) 239.44
MDL Number MFCD00661298
InChI Key OHWSWGXNZDSHLM-UHFFFAOYSA-N
Synonym 2-chloro-3-iodo-pyridine, pyridine, 2-chloro-3-iodo, 3-iodo-2-chloropyridine, zlchem 306, pubchem1188, acmc-209pes, 2-chloro-3-iodopyridine, ksc378k0b, 2-chloranyl-3-iodanyl-pyridine, 2-choro-3-iodopyridine
PubChem CID 2735743
IUPAC Name 2-chloro-3-iodopyridine
SMILES C1=CC(=C(N=C1)Cl)I
Specifications
Melting Point 93°C to 97°C
Quantity 1 g
Beilstein 4243304
Sensitivity Light sensitive
Formula Weight 239.44
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-iodopyridine