Learn More
2-Chloro-1,5-hexadiene (>90%), TRC
Product Code. 30368323
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
missing translation for 'unitSize':
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30368323 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Product Code. 30368323 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC364010100MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloro-1,5-hexadiene
CAS 101933-88-4
Molecular Formula C6H9Cl
Quantity 100 mg
Synonym 2-Chlorohexa-1,5-Diene
InChI Formula InChI=1S/C6H9Cl/c1-3-4-5-6(2)7/h3H,1-2,4-5H2
SMILES ClC(CCC=C)=C
IUPAC Name 2-Chloro-1,5-hexadiene (ACI)
Molecular Weight (g/mol) 116.59
Formula Weight 116.04