Learn More
2-Chloro-1,3-thiazole, ≥97%, Thermo Scientific™
Product Code. p-3175015
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
Product Code. 10215794
Brand: Thermo Scientific Maybridge MO07794CB
Request Bulk or Custom Format
Chemical Identifiers
CAS 3034-52-4
Molecular Formula C3H2ClNS
Molecular Weight (g/mol) 119.566
MDL Number MFCD00210701
InChI Key KLEYVGWAORGTIT-UHFFFAOYSA-N
Synonym 2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569
PubChem CID 76429
ChEBI CHEBI:39187
IUPAC Name 2-chloro-1,3-thiazole
SMILES C1=CSC(=N1)Cl
Specifications
CAS Min % 97.0
Density 1.3780g/mL
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 119.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-1, 3-thiazole