missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate, 98%, Thermo Scientific™

Product Code. 15415076
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415076 1 g 1g
15425076 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415076 Supplier Thermo Scientific Alfa Aesar Supplier No. H59515.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101385-69-7
Molecular Formula C5H10ClF6N2P
Molecular Weight (g/mol) 278.563
MDL Number MFCD00191914
InChI Key CNAKHAGVVMOXFE-UHFFFAOYSA-N
Synonym 2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium hexafluorophosphate v, 2-chloro-1,3-dimethylimidazolinium hexafluorophosphate, 2-chloro-1,3-dimethylimidazolidium hexafluorophosphate, 2-chloro-1,3-dimethylimidazolidinium hexafluorophosphate, cip, 2-chloro-1,3-dimethylimidazolidinium hexafluorophosphate, 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium hexafluorophosphate, 2-chloro-4,5-dihydro-1,3-dimethyl-1h-imidazolium hexafluorophosphate, 2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium; hexafluoro-lambda5-phosphanuide, 2-chloro-1,3-dimethylimidazolidinium hexafluorophosphate, purum t
PubChem CID 16211675
IUPAC Name 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium;hexafluorophosphate
SMILES CN1CC[N+](=C1Cl)C.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 231°C to 233°C
Quantity 1 g
Beilstein 5369058
Sensitivity Light and moisture sensitive
Formula Weight 278.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.